Merck
  • Reaction pathways of the diketonitrile degradate of isoxaflutole with hypochlorite in water.
Reaction pathways of the diketonitrile degradate of isoxaflutole with hypochlorite in water.
Journal of agricultural and food chemistry (2007-02-08)
R N Lerch, C H Lin, N D Leigh
ABSTRACT
Isoxaflutole (IXF; Balance) belongs to a new class of isoxazole herbicides. Isoxaflutole has a very short half-life in soil and rapidly degrades to a stable and phytotoxic degradate, diketonitrile (DKN). DKN was previously discovered to rapidly react with hypochlorite (OCl-) in tap water, yielding the benzoic acid (BA) degradate as a major product, but the complete reaction pathway and mechanism have not been elucidated. Thus, the objectives of this work were to (1) determine the stoichiometry of the reaction between DKN and OCl-; (2) identify products in addition to BA; and (3) propose a complete pathway and reaction mechanism for oxidation of DKN by OCl-. Stoichiometry of the reaction showed a molar ratio of OCl-/DKN of 2. In addition, two previously uncharacterized chlorinated intermediates were identified under conditions in which OCl- was the limiting reactant. The proposed chemical structure of a chlorinated benzoyl intermediate was inferred from a series of HPLC/MS and HPLC/MS/MS experiments and the use of mass spectral simulation software. A chlorinated ketone intermediate was also identified using ion trap GC/MS. Two additional end products were also identified: cyclopropanecarboxylic acid (CPCA) and dichloroacetonitrile (DCAN). On the basis of the reaction stoichiometry, the structure of the chlorinated intermediates, and the identification of the products, two reaction pathways are proposed. Both pathways involve a two-step nucleophilic attack and oxidation of the diketone structure of DKN, leading to formation of BA, DCAN, and CPCA.
Product Description
Supelco
Isoxaflutole, PESTANAL®, analytical standard