Merck
  • Cu-catalyzed oxidative amidation of propiolic acids under air via decarboxylative coupling.
Cu-catalyzed oxidative amidation of propiolic acids under air via decarboxylative coupling.
Organic letters (2010-04-07)
Wei Jia, Ning Jiao
ABSTRACT
A Cu-catalyzed aerobic oxidative amidation of propiolic acids via decarboxylation under air has been developed. Only carbon dioxide is produced as byproduct in this approach. The use of air as oxidant makes this method more useful and easy to handle.
Sigma-Aldrich
Propiolic acid, 95%