Merck
What happened to WHI: menopausal hormonal therapy in 2012.
Clinical obstetrics and gynecology (2012-07-26)
Keith A Hansen, Kathleen M Eyster
ABSTRACT
Menopause is characterized by amenorrhea for 1 year due to the cessation of ovarian function. The hormonal treatment of menopause has significantly altered since the publication of initial results from the Women's Health Initiative continuous, combined, conjugated equine estrogen with medroxyprogesterone acetate study arm in 2002. Current studies suggest that treatment should be individualized and that the lowest dose of estrogen providing relief should be used for the shortest period of time in menopausal women who experience vasomotor symptoms or urogenital atrophy. Future studies into different delivery mechanisms such as transdermal applications and different agents, such as tibolone and raloxifene, will help refine the treatment of menopause.
Product Description
Tibolone for system suitability, European Pharmacopoeia (EP) Reference Standard