Merck
  • A comparative study of buffer systems as cryoprotectants for human spermatozoa.
A comparative study of buffer systems as cryoprotectants for human spermatozoa.
Fertility and sterility (1986-07-01)
G S Prins, L Weidel
ABSTRACT
Several cryoprotective media currently in use for human sperm cryopreservation have been directly compared with each other by use of a static vapor freezing technique. The TEST-citrate-yolk buffers with glycerol resulted in the highest motility and longevity after thawing; thus, their use is recommended.
Sigma-Aldrich
TES, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
TES, ≥99% (titration)
Sigma-Aldrich
TES, BioXtra, ≥99.5% (titration)