Merck
  • Band composition analysis: a new procedure for deconvolution of the mass spectra of organometallic compounds.
Band composition analysis: a new procedure for deconvolution of the mass spectra of organometallic compounds.
Journal of mass spectrometry : JMS (2003-08-26)
Jacek A Szymura, Jan Lamkiewicz
ABSTRACT
A new chemometric procedure called band composition analysis (BCA) designed for the deconvolution of mass spectra of organometallics is proposed. BCA generates theoretical bands T(i), then combines them to obtain a model band M, which is finally compared with the experimental band E. All of these steps are realized with computer assistance. This modeling yields four parameters characterizing the experimental band: theoretical and model variances s(2) (theor) and s(2) (model), a fit factor alpha and a contribution x(i) from the theoretical band. If s(2) (theor) > 20 the band is deemed complex and needs modeling. The values alpha > 90 indicate that there is good agreement between the experimental and model bands. BCA is particularly effective for the modeling of complex isotopic bands often present in organometallics. Two illustrations of BCA for tetrabutyltin, C(16)H(36)Sn, and 1,1',2,2',3,3'-hexachloroferrocene, C(10)H(4)Cl(6)Fe, are shown.
Sigma-Aldrich
Tetrabutyltin, technical grade, 93%