Merck
  • Fucoxanthin: a marine carotenoid exerting anti-cancer effects by affecting multiple mechanisms.
Fucoxanthin: a marine carotenoid exerting anti-cancer effects by affecting multiple mechanisms.
Marine drugs (2013-12-20)
Sangeetha Ravi Kumar, Masashi Hosokawa, Kazuo Miyashita
ABSTRACT
Fucoxanthin is a marine carotenoid exhibiting several health benefits. The anti-cancer effect of fucoxanthin and its deacetylated metabolite, fucoxanthinol, is well documented. In view of its potent anti-carcinogenic activity, the need to understand the underlying mechanisms has gained prominence. Towards achieving this goal, several researchers have carried out studies in various cell lines and in vivo and have deciphered that fucoxanthin exerts its anti-proliferative and cancer preventing influence via different molecules and pathways including the Bcl-2 proteins, MAPK, NFκB, Caspases, GADD45, and several other molecules that are involved in either cell cycle arrest, apoptosis, or metastasis. Thus, in addition to decreasing the frequency of occurrence and growth of tumours, fucoxanthin has a cytotoxic effect on cancer cells. Some studies show that this effect is selective, i.e., fucoxanthin has the capability to target cancer cells only, leaving normal physiological cells unaffected/less affected. Hence, fucoxanthin and its metabolites show great promise as chemotherapeutic agents in cancer.
Product Description
Supelco
Fucoxanthin, analytical standard
Sigma-Aldrich
Fucoxanthin, carotenoid antioxidant