Merck
  • Body mass index in chronic heart failure: association with biomarkers of neurohormonal activation, inflammation and endothelial dysfunction.
Body mass index in chronic heart failure: association with biomarkers of neurohormonal activation, inflammation and endothelial dysfunction.
BMC cardiovascular disorders (2013-10-03)
Heidi M Christensen, Morten Schou, Jens P Goetze, Jens Faber, Jan Frystyk, Allan Flyvbjerg, Caroline Kistorp
ABSTRACT
Low body mass index (BMI) is associated with a poor outcome in chronic heart failure (CHF). An inverse association between BMI and adiponectin and N-terminal pro-B-type natriuretic peptide (NT-proBNP) has been reported. The aim of the present study was to investigate whether novel markers of neurohormonal activation, inflammation, and endothelial dysfunction are associated with BMI in CHF. In a cross-sectional study including 171 patients with CHF and a left ventricular ejection fraction (LVEF) ≤45% the impact of BMI on circulating plasma concentrations of adiponectin, α-defensins, high sensitivity C-reactive protein (hsCRP), copeptin, mid-regional pro-adrenomedullin (MR-proADM), NT-proBNP, and mid-regional pro-A-type natriuretic peptide (MR-proANP) were evaluated. In multivariable linear regression analysis including age, sex, LVEF, New York Heart Association functional classification (NYHA), estimated glomerular filtration rate (eGFR), and diabetes, only NT-proBNP (β = -0.32) and adiponectin (β = -0.39) remained independently associated with BMI. MR-proANP was associated with BMI but adjusting for age attenuated the relation being no longer significant. Among biomarkers typically increased in patients with CHF only adiponectin and NT-proBNP demonstrated independent inverse associations with BMI. This indicates a direct effect of these two biomarkers enhancing the wasting process seen in CHF.
Sigma-Aldrich
Adiponectin human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)