
Patchouli alcohol

Synonym(s):
(1R,3R,6S)-2,2,6,8-Tetramethyltricyclo[5.3.1.03,8]undec-3-ol
Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1