Skip to Content
Merck
Rinse Fluid A bottle of 1 L, pkg of 4 units, square polycarbonate bottle (1L with Standard Cap), sterile; autoclaved

Rinse Fluid A

Rinse Fluid A bottle of 1 L, pkg of 4 units, square polycarbonate bottle (1L with Standard Cap), sterile; autoclaved

Products

Product Category
Physical Form
Agency
Quality Segment
Application
Bottle material
Vial Bottle Material
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique