
Valerian standardised dry extract

Synonym(s):
Valerenic acid, (2E)-3-[(4S,7R,7aR)-2,4,5,6,7,7a-Hexadydro-3,7-dimethyl- 1H-inden-4-yl]-2-methyl-2-propenoic acid
Empirical Formula (Hill Notation):
C15H22O2
CAS Number:
Molecular Weight:
234.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing