Merck
  • The Influence of Chemical Structure and the Presence of Ascorbic Acid on Anthocyanins Stability and Spectral Properties in Purified Model Systems.
The Influence of Chemical Structure and the Presence of Ascorbic Acid on Anthocyanins Stability and Spectral Properties in Purified Model Systems.
Foods (Basel, Switzerland) (2019-06-20)
Rachel Levy, Zoya Okun, Avi Shpigelman
ABSTRACT
The loss of color pigment is an important quality factor of food products. This work aimed to systematically study, in purified model systems, the influence of anthocyanins' structure (by increasing the size of the conjugated sugar) and the presence of ascorbic acid on their stability and spectral properties during storage at two pH levels relevant to medium and high acid foods (6.5 and 4.5, respectively). Anthocyanins (cyanidin (Cy), cyanidin 3-O-β-glucoside (Cy3G) and cyanidin 3-O-β-rutinoside (Cy3R)) displayed first-order degradation rates, presenting higher stability in acidic medium and enhanced stability with increasing size of conjugated sugar. The addition of ascorbic acid resulted in significantly enhanced degradation. Changes in ultra violet visible (UV-VIS) spectral properties presented a decrease in typical color intensity and pointed towards formation of degradation products. Identification and kinetics of formation for cyanidin degradation products were obtained by high performance liquid chromatography system-mass spectrometry (HPLC-MS).