Merck
  • Early goal-directed haemodynamic optimization of cerebral oxygenation in comatose survivors after cardiac arrest: the Neuroprotect post-cardiac arrest trial.
Early goal-directed haemodynamic optimization of cerebral oxygenation in comatose survivors after cardiac arrest: the Neuroprotect post-cardiac arrest trial.
European heart journal (2019-03-22)
Koen Ameloot, Cathy De Deyne, Ward Eertmans, Bert Ferdinande, Matthias Dupont, Pieter-Jan Palmers, Tibaut Petit, Philippe Nuyens, Joren Maeremans, Joris Vundelinckx, Maarten Vanhaverbeke, Ann Belmans, Ronald Peeters, Philippe Demaerel, Robin Lemmens, Jo Dens, Stefan Janssens
ABSTRACT
During the first 6-12 h of intensive care unit (ICU) stay, post-cardiac arrest (CA) patients treated with a mean arterial pressure (MAP) 65 mmHg target experience a drop of the cerebral oxygenation that may cause additional cerebral damage. Therefore, we investigated whether an early goal directed haemodynamic optimization strategy (EGDHO) (MAP 85-100 mmHg, SVO2 65-75%) is safe and could improve cerebral oxygenation, reduce anoxic brain damage, and improve outcome when compared with a MAP 65 mmHg strategy. A total of 112 out-of-hospital CA patients were randomly assigned to EGDHO or MAP 65 mmHg strategies during the first 36 h of ICU stay. The primary outcome was the extent of anoxic brain damage as quantified by the percentage of voxels below an apparent diffusion coefficient (ADC) score of 650.10-6 mm2/s on diffusion weighted magnetic resonance imaging (at day 5 ± 2 post-CA). Main secondary outcome was favourable neurological outcome (CPC score 1-2) at 180 days. In patients assigned to EGDHO, MAP (P < 0.001), and cerebral oxygenation during the first 12 h of ICU stay (P = 0.04) were higher. However, the percentage of voxels below an ADC score of 650.10-6 mm2/s did not differ between both groups [16% vs. 12%, odds ratio 1.37, 95% confidence interval (CI) 0.95-0.98; P = 0.09]. Also, the number of patients with favourable neurological outcome at 180 days was similar (40% vs. 38%, odds ratio 0.98, 95% CI 0.41-2.33; P = 0.96). The number of serious adverse events was lower in patients assigned to EGDHO (P = 0.02). Targeting a higher MAP in post-CA patients was safe and improved cerebral oxygenation but did not improve the extent of anoxic brain damage or neurological outcome.
Sigma-Aldrich
2-Methyl-2-propanethiol, 99%