Merck
  • Interaction of Commonly Used Oral Molecular Excipients with P-glycoprotein.
Interaction of Commonly Used Oral Molecular Excipients with P-glycoprotein.
The AAPS journal (2021-09-17)
Ruchika Bajaj, Lisa B Chong, Ling Zou, Eleftheria Tsakalozou, Zhanglin Ni, Kathleen M Giacomini, Deanna L Kroetz
ABSTRACT
P-glycoprotein (P-gp) plays a critical role in drug oral bioavailability, and modulation of this transporter can alter the safety and/or efficacy profile of substrate drugs. Individual oral molecular excipients that inhibit P-gp function have been considered a mechanism for improving drug absorption, but a systematic evaluation of the interaction of excipients with P-gp is critical for informed selection of optimal formulations of proprietary and generic drug products. A library of 123 oral molecular excipients was screened for their ability to inhibit P-gp in two orthogonal cell-based assays. β-Cyclodextrin and light green SF yellowish were identified as modest inhibitors of P-gp with IC50 values of 168 μM (95% CI, 118-251 μM) and 204 μM (95% CI, 5.9-1745 μM), respectively. The lack of effect of most of the tested excipients on P-gp transport provides a wide selection of excipients for inclusion in oral formulations with minimal risk of influencing the oral bioavailability of P-gp substrates.
Sigma-Aldrich
Elacridar, ≥98% (HPLC)