Merck
  • Synthesis and antimicrobial activities of two novel amino sugars derived from chloraloses.
Synthesis and antimicrobial activities of two novel amino sugars derived from chloraloses.
Carbohydrate research (2010-05-22)
Nilgün Yenil, Emriye Ay, Kadir Ay, Mustafa Oskay, Jacques Maddaluno
ABSTRACT
The synthesis of 5-amino-5-deoxy-1,2-O-(S)-trichloroethylidene-beta-L-arabinofuranose and 6-amino-6-deoxy-1,2-O-(S)-trichloroethylidene-alpha-D-glucofuranose is described by a simple three- or four-step route. Antibacterial potency of the new compounds was determined using an inhibition zone diameter test. The results show that these compounds have a broad-spectrum activity against Gram-positive, Gram-negative bacteria and Candida albicans.
Sigma-Aldrich
α-Chloralose, ≥98.0% (chloralose, AT)
Sigma-Aldrich
α-Chloralose
Supelco
Chloralose, PESTANAL®, analytical standard