Merck
  • Synergistic antibacterial brilliant blue/reduced graphene oxide/quaternary phosphonium salt composite with excellent water solubility and specific targeting capability.
Synergistic antibacterial brilliant blue/reduced graphene oxide/quaternary phosphonium salt composite with excellent water solubility and specific targeting capability.
Langmuir : the ACS journal of surfaces and colloids (2011-05-19)
Xiang Cai, Shaozao Tan, Minsong Lin, Agui Xie, Wenjie Mai, Xiuju Zhang, Zhidan Lin, Ting Wu, Yingliang Liu
ABSTRACT
A water-soluble brilliant blue/reduced graphene oxide/tetradecyltriphenylphosphonium bromide composite (BB-rGO-TTP) was prepared by using noncovalent brilliant blue-functionalized reduced graphene oxide (BB-rGO) as the tetradecyltriphenylphosphonium bromide (TTP) carrier. Antibacterial performance of this novel composite was investigated for both Gram-positive and Gram-negative bacteria. The results showed that the novel BB-rGO-TTP, combing the advantages of graphene and TTP, displayed excellent synergistic antibacterial activity, specific targeting capability, water solubility, and mild cytotoxicity, suggesting the great potential application as sprayable graphene-based antibacterial solutions.
Sigma-Aldrich
Erioglaucine disodium salt, coloring dye