![[(IMes)AuCl] Umicore](/deepweb/assets/sigmaaldrich/product/structures/154/609/370330fe-5c15-47b6-ad13-ea3dc87099d6/640/370330fe-5c15-47b6-ad13-ea3dc87099d6.png)
[(IMes)AuCl]
![[(IMes)AuCl] Umicore](/deepweb/assets/sigmaaldrich/product/structures/154/609/370330fe-5c15-47b6-ad13-ea3dc87099d6/640/370330fe-5c15-47b6-ad13-ea3dc87099d6.png)
Synonym(s):
Chloro[1,3-bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]gold(I), 1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene gold(I) chloride
Empirical Formula (Hill Notation):
C21H24AuClN2
CAS Number:
Molecular Weight:
536.85
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing