Merck
Neutral gold complexes with tridentate SNS thiosemicarbazide ligands.
Inorganic chemistry (2012-01-12)
Pedro I da S Maia, Hung Huy Nguyen, Daniela Ponader, Adelheid Hagenbach, Silke Bergemann, Ronald Gust, Victor M Deflon, Ulrich Abram
ABSTRACT
Na[AuCl(4)]·2H(2)O reacts with tridentate thiosemicarbazide ligands, H(2)L1, derived from N-[N',N'-dialkylamino(thiocarbonyl)]benzimidoyl chloride and thiosemicarbazides under formation of air-stable, green [AuCl(L1)] complexes. The organic ligands coordinate in a planar SNS coordination mode. Small amounts of gold(I) complexes of the composition [AuCl(L3)] are formed as side-products, where L3 is an S-bonded 5-diethylamino-3-phenyl-1-thiocarbamoyl-1,2,4-triazole. The formation of the triazole L3 can be explained by the oxidation of H(2)L1 to an intermediate thiatriazine L2 by Au(3+), followed by a desulfurization reaction with ring contraction. The chloro ligands in the [AuCl(L1)] complexes can readily be replaced by other monoanionic ligands such as SCN(-) or CN(-) giving [Au(SCN)(L1)] or [Au(CN)(L1)] complexes. The complexes described in this paper represent the first examples of fully characterized neutral Gold(III) thiosemicarbazone complexes. All the [AuCl(L1)] compounds present a remarkable cell growth inhibition against human MCF-7 breast cancer cells. However, systematic variation of the alkyl groups in the N(4)-position of the thiosemicarbazone building blocks as well as the replacement of the chloride by thiocyanate ligands do not considerably influence the biological activity. On the other hand, the reduction of Au(III) to Au(I) leads to a considerable decrease of the cytotoxicity.
Sigma-Aldrich
Thiosemicarbazide, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
1 Estimated to ship on 19 March 2025
Details...
£139.00
1 kg
1 Estimated to ship on 06 January 2025
Details...
£258.00
Sigma-Aldrich
Thiosemicarbazide, puriss. p.a., 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
1 Estimated to ship on 19 March 2025
Details...
£139.00
1 kg
1 Estimated to ship on 06 January 2025
Details...
£258.00