Merck
  • Effect of genetic polymorphism of UCP2-866 G/A on repaglinide response in Chinese patients with type 2 diabetes.
Effect of genetic polymorphism of UCP2-866 G/A on repaglinide response in Chinese patients with type 2 diabetes.
Die Pharmazie (2012-03-08)
Shan Wang, Yan-Mei Se, Zhao-Qian Liu, Ming-Xiang Lei, Hao-BoYang, Zhi-Xiang Sun, Sheng-Dan Nie, Xiao-min Zeng, Jing Wu
ABSTRACT
The aim of the present study was to evaluate the impact of the UCP2-866 G/A polymorphism on the efficacy of repaglinide in treating patients with diabetes mellitus type 2 (T2DM). 370 patients with T2DM and 166 healthy volunteers were enrolled to identify UCP2-866 G/A genotypes. 16 patients with GG genotype, 14 with GA genotype and 11 with AA genotype of UCP2-866 G/A underwent an 8-week repaglinide treatment regimen. There were no differences in allele frequency of UCP2-866 G/A between T2DM patients and control subjects. The patient with AA genotype of UCP2-866 G/A had higher levels of fasting plasma glucose (FPG), 30-min and 2-h postload plasma glucose, glycated haemoglobin (HbA1c), and lower concentrations of 30-min and 2-h postload plasma insulin, homeostasis model assessment of beta cell function (HOMA-beta), deltaI30/deltaG30 compared with GG genotype. After repaglinide treatment for 8 consecutive weeks, we found that A allele carriers of UCP2 in the T2DM patients had smaller decrease in FPG (P < 0.05) and HbA1c (P < 0.05), and smaller increase in 30-min postload plasma insulin (P < 0.01) compared with GG genotypes. We demonstrated that UCP2-866 G/A polymorphism is associated with the therapeutic efficacy of repaglinide in Chinese T2DM patients.
Product Description
Repaglinide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Repaglinide, ≥98% (HPLC), solid
Repaglinide, European Pharmacopoeia (EP) Reference Standard