Merck
  • NTP, the photoproduct of nifedipine, activates caffeine-sensitive ion channels in leech neurons.
NTP, the photoproduct of nifedipine, activates caffeine-sensitive ion channels in leech neurons.
Cell calcium (2003-02-26)
Jürgen Schoppe, Paul W Dierkes, Peter Hochstrate, Wolf R Schlue
ABSTRACT
Leech P neurons possess caffeine-sensitive ion channels in intracellular Ca(2+) stores and in the plasma membrane. The following results indicate that these channels are also activated by 2,6-dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester (NTP), the photoproduct of the L-type Ca(2+) channel-blocker nifedipine: (1) Just like caffeine, NTP evoked Ca(2+) influx and intracellular Ca(2+) release, as well as the influx of various other divalent cations and that of Na(+). (2) In the presence of high NTP or caffeine concentrations the plasma membrane channels close, suggesting desensitization of the channel-activating mechanism. (3) Depending on the concentration, NTP and caffeine induce cross-desensitization or act additively. (4) NTP was effective in the same neurons as caffeine (P, N, Leydig, 101), and it was ineffective in neurons in which caffeine was also ineffective (AP, T, L, 8, AE). (5) In Retzius neurons, NTP and caffeine evoked intracellular Ca(2+) release but no Ca(2+) influx. Despite these parallels, the effects of NTP and caffeine were not identical, which may be due to differences in the mechanisms of channel activation or desensitization and/or to substance-specific side effects. The caffeine-sensitive ion channels were activated by NTP concentrations > or =10 microM, which is almost three orders of magnitude smaller than the threshold concentration of caffeine.
Product Description
USP
Nifedipine Nitrosophenylpyridine Analog, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Nifedipine impurity B, European Pharmacopoeia (EP) Reference Standard