Skip to Content
Merck
2,6-Dimethoxyphenol analytical standard

2,6-Dimethoxyphenol

2,6-Dimethoxyphenol analytical standard
Synonym(s):
Pyrogallol 1,3-dimethyl ether
Linear Formula:
(CH3O)2C6H3OH
CAS Number:
Molecular Weight:
154.16
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing