Deoxo-Fluor solution
Synonym(s):
Bis(2-methoxyethyl)aminosulfur trifluoride solution
Empirical Formula (Hill Notation):
C6H14F3NO2S
CAS Number:
Molecular Weight:
221.24
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing