
DMAT

Synonym(s):
CK2 Inhibitor II, 2-Dimethylamino-4,5,6,7-tetrabromo-1H-benzimidazole, DMAT
Empirical Formula (Hill Notation):
C9H7Br4N3
CAS Number:
Molecular Weight:
476.79
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing