
Nafcillin sodium salt

Synonym(s):
(2S,5R,6R)-6-{[(2-Ethoxy-1-naphthalenyl)carbonyl]amino}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium salt, 6-(2-Ethoxy-1-naphthamido)penicillin sodium salt
Empirical Formula (Hill Notation):
C21H21N2NaO5S
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing