
Nilotinib impurity A

Synonym(s):
3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline
Empirical Formula (Hill Notation):
C11H10F3N3
CAS Number:
Molecular Weight:
241.21
Products
Formula Weight
Manufacturer
Format
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing