
SP Brij® S2 MBAL

Synonym(s):
Brij® S2, Polyethylene glycol octadecyl ether, Polyoxyethylene (2) stearyl ether
Linear Formula:
C18H37(OCH2CH2)2OH
CAS Number:
Molecular Weight:
358.60
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing