Merck
  • Lasting alterations induced in glial cell phenotypes by short exposure to alcohol during embryonic development in zebrafish.
Lasting alterations induced in glial cell phenotypes by short exposure to alcohol during embryonic development in zebrafish.
Addiction biology (2020-01-11)
Dipashree Chatterjee, Samantha Mahabir, Diptendu Chatterjee, Robert Gerlai
ABSTRACT
Despite the known teratogenic effects of alcohol (ethanol) on the developing human fetus, the prevalence of fetal alcohol spectrum disorder (FASD) is not decreasing. Appropriate treatment for this life-long disease has not been developed, and even diagnostic biomarkers are unavailable. FASD remains a large unmet medical need. Numerous animal models have been developed to mimic FASD and study potential underlying biological mechanisms. However, most of these models focused on neuronal phenotypes. Given that glial cells represent the majority of cells in the vertebrate brain, and given the increasingly appreciated roles they play in a myriad of neuronal functions as well as CNS disorders, we decided to investigate potential embryonic alcohol exposure induced changes in them. Building upon a previously introduced zebrafish model of milder and most prevalent forms of FASD, we investigated the effect of a 2-hour-long exposure to alcohol (1% vol/vol bath concentration) employed at the 24th hour postfertilization stage of development of zebrafish on a number of glial cell-related phenotypes. We studied oligodendrocyte, astrocyte as well as microglia-related phenotypes using immunohistochemistry, lipid, and enzyme activity analyses. We report significant changes in wide-spread glial cell phenotypes induced by embryonic alcohol exposure in the zebrafish brain and conclude that the zebrafish will advance our understanding of the mechanisms of this devastating disorder.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−TRITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Galactocerebrosides from bovine brain, ≥97% (TLC)
Sigma-Aldrich
Cholera Toxin B subunit, FITC conjugate, lyophilized powder
Sigma-Aldrich
Anti-Galactocerebroside antibody produced in rabbit, whole antiserum