Merck
Copper inclusion in cellulose using sodium D-gluconate complexes.
Carbohydrate polymers (2012-09-04)
Hossam E Emam, Avinash P Manian, Barbora Siroká, Thomas Bechtold
ABSTRACT
Copper containing cellulose material is of growing interest, e.g. offering alternative in the field of antimicrobials. Solutions of copper d-gluconate complexes (Cu(2+)-DGL) were used to introduce copper ions into a swollen cellulosic matrix. A ligand exchange mechanism forms the chemical basis of the sorption process. Copper sorption in cellulose was studied in the range between pH 6 and 13. An estimate for the complex stabilities of the Cu-cellulose system could be derived from the calculated species distribution of the different Cu(2+)-DGL complexes present. Spectrophotometry and cyclic voltammetry of Cu(2+)-DGL complex solution were used to confirm the presence of different species participating in the ligand exchange reaction. The pH dependent uptake of Cu(2+) ions in the cellulose matrix can be explained on the basis of the relative stabilities of Cu(2+)-DGL complex vs. Cu(2+)-cellulose complexes. In comparison to pH 10, higher copper content was observed at pH 6 and 13. Copper content was limited by carboxyl content of cellulosic materials, thus in analogy to the structure of Cu(2+)-DGL complexes participation of the carboxyl group as complex forming site is proposed. At high Cu(2+)-concentration and longer time of immersion in the copper complex solutions formation of solid deposits was observed on the surface of the treated fibres.
Sigma-Aldrich
Sodium gluconate, meets USP testing specifications
Supelco
D-Gluconic acid sodium salt, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-Gluconic acid sodium salt, ≥99% (HPLC)
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Sigma-Aldrich
Magnesium D-gluconate hydrate, 98.0-102% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(II) D-gluconate dihydrate, 98%
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Magnesium D-gluconate hydrate, ≥98% (HPLC)