Merck
  • Contact allergy to the UV absorber Tinuvin P in a dental restorative material.
Contact allergy to the UV absorber Tinuvin P in a dental restorative material.
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society (1997-03-01)
B Björkner, B Niklasson
ABSTRACT
A case of contact allergy to the UV absorber Tinuvin P is described in a patient with gingivitis who was previously treated with a dental restorative material. With high-performance liquid chromatography analysis, the presence of Tinuvin P could be shown in the restorative material.
Sigma-Aldrich
2-(2-Hydroxy-5-methylphenyl)benzotriazole, 97%