Merck
  • Quinone mixture as attractant for necrophagous dung beetles specialized on dead millipedes.
Quinone mixture as attractant for necrophagous dung beetles specialized on dead millipedes.
Journal of chemical ecology (2004-07-21)
Thomas Schmitt, Frank-Thorsten Krell, K Eduard Linsenmair
ABSTRACT
2-Methyl-1,4-benzoquinone (toluquinone) and 2-methoxy-3-methyl-1,4-benzoquinone are the most common components of defensive secretions of juliform millipedes (Diplopoda: Juliformia). A natural and a synthetic millipede-defensive secretion composed of these two substances attract dung beetles of a few Onthophagus species (Coleoptera: Scarabaeidae) that feed mainly on freshly dead millipedes. This olfactory mechanism and adaptation to the toxic effects of quinones enables them to be the first and exclusive users of this resource.
Sigma-Aldrich
Methyl-p-benzoquinone, 98%