Merck
  • The major metabolite of dipeptide piracetam analogue GVS-111 in rat brain and its similarity to endogenous neuropeptide cyclo-L-prolylglycine.
The major metabolite of dipeptide piracetam analogue GVS-111 in rat brain and its similarity to endogenous neuropeptide cyclo-L-prolylglycine.
European journal of drug metabolism and pharmacokinetics (1997-07-01)
T A Gudasheva, S S Boyko, R U Ostrovskaya, T A Voronina, V K Akparov, S S Trofimov, G G Rozantsev, A P Skoldinov, V P Zherdev, S B Seredenin
ABSTRACT
The metabolism of a new piracetam analogue, the dipeptide cognitive enhancer N-phenylacetyl-L-prolylglycine ethyl ester (GVS-111) was studied in vivo. GVS-111 itself was not found in rat brain 1 h after 5 mg/kg i.p. administration up to limit of detection (LOD) under high performance liquid chromatography (HPLC) conditions. Three substances corresponding to the three possible GVS-111 metabolites, namely phenylacetic acid, prolylglycine and cyclo-prolylglycine, were found in experimental rat brain samples as well as in controls using HPLC, gas chromatography (GC) and gas chromatography-mass spectrometry (GC-MS) methods. Only cyclo-prolylglycine concentration increased (2.5-fold) 1 h after GVS-111 administration. Cyclo-prolylglycine formation from GVS-111 in the presence of plasma and brain enzymes was shown in vitro. These data could be considered as evidence that GVS-111 is prodrug which converts in the body to cyclo-prolylglycine, and which is identical to the endogenous cyclopeptide that produces the nootropic activity.
Sigma-Aldrich
Pro-Gly