Merck
  • Supplementing rooster sperm with Cholesterol-Loaded-Cyclodextrin improves fertility after cryopreservation.
Supplementing rooster sperm with Cholesterol-Loaded-Cyclodextrin improves fertility after cryopreservation.
Cryobiology (2017-01-04)
Napapach Chuaychu-Noo, Pachara Thananurak, Vibantita Chankitisakul, Thevin Vongpralub
ABSTRACT
Little is known about the effects of Cholesterol-Loaded Cyclodextrin (CLC) on post-thaw semen quality in chicken. The aim of the present study is to investigate the efficacy of CLC levels (0, 1, 2 and 3 mg/mL Schramm diluent) on post-thawed semen quality and fertility in two breeds of chicken Pradu Hang Dum (native chicken) and Rhode Island Red. Semen samples of each breed were pooled, divided into 4 aliquots and diluted with Schramm diluents, cooled to 5 °C when DMF was added (6% of final volume). Semen straws were subjected to cryopreservation using the liquid nitrogen vapor method. Post-thawed sperm motility, viability, acrosome integrity, mitochondrial function, and the Malondialdehyde (MDA) level were determined. The fertility of frozen semen was tested by inseminating laying hens. Post-thaw motility between Pradu Hang Dum and Rhode Island Red was no different; but Rhode Island Red had a higher semen viability and live cell intact acrosomes than Pradu Hang Dum (P < 0.05). The percentage of high functioning mitochondria in the Pradu Hang Dum was higher than the Rhode Island Red. CLC at 2 and 3 mg/mL supplementation was associated with improved viability of frozen semen; that is, acrosome integrity and mitochondrial function (P < 0.01), albeit having no effect on MDA levels. The sperm with 1 mg/mL CLC yielded a significantly better fertility (P < 0.01). CLC (1 mg/mL) improved the quality of frozen rooster semen. There was no interaction among breeds and CLC on post-thaw semen quality and fertility.
Sigma-Aldrich
Lectin from Arachis hypogaea (peanut), FITC conjugate, lyophilized powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2 tabs
Estimated to ship on 28 February 2025
from MILWAUKEE
$66.50
20 tabs
Estimated to ship on 28 February 2025
from MILWAUKEE
$510.00