Merck
Effects of arginine and trehalose on post-thawed bovine sperm quality.
Acta veterinaria Hungarica (2017-09-29)
Caner Öztürk, Şükrü Güngör, Mehmet Bozkurt Ataman, Mustafa Numan Bucak, Nuri Başpinar, Pınar Ili, Muhammed Enes Inanç
ABSTRACT
The present study was conducted to examine the protective role of arginine and trehalose on post-thaw bull sperm and oxidative stress parameters. Five ejaculates for each bull were used in the study. Each ejaculate, split into three equal aliquots and diluted at 37 °C with base extenders containing 2 mM arginine, 25 mM trehalose and no antioxidant (control) was cooled to 5 °C and then frozen. Frozen straws were thawed in a water bath for evaluation. Supplementation of the semen extender with arginine decreased the percentages of post-thawed subjective motility (29 ± 8.21%), CASA motility (12.2 ± 5.69%) and progressive motility (3.52 ± 2.13%), compared with the controls (43 ± 2.73%, 55.4 ± 6.78% and 33.48 ± 4.14%, respectively, P < 0.05). Supplementation of the semen extender with trehalose produced a higher mitochondrial activity and sperm viability (36.3 ± 3.99% and 44.1 ± 2.18%) compared with the control (13 ± 8.15 and 31.7 ± 3.94%, respectively, P < 0.05). It was established that trehalose (95.1%) and arginine (92.8%) protect DNA integrity compared to the control (90.4%) (P < 0.05). Trehalose supplementation in semen extenders provided great benefit in terms of viability, mitochondrial activity, and intact sperm DNA on frozen-thawed bull sperm.
Sigma-Aldrich
Lectin from Arachis hypogaea (peanut), FITC conjugate, lyophilized powder