- Suramin Affects the Renal VEGF-A/VEGFR Axis in Short-Term Streptozotocin-Induced Diabetes.
Suramin Affects the Renal VEGF-A/VEGFR Axis in Short-Term Streptozotocin-Induced Diabetes.
Diabetic nephropathy (DN) accounts for approximately 50% of end-stage renal diseases. Vascular endothelial growth factor A (VEGF-A) is thought to be a critical mediator of vascular dysfunction in DN, but its role is unclear. The lack of pharmacological tools to modify renal concentrations further hinders the understanding of its role in DN. In this study, rats were evaluated after 3 weeks of streptozotocin-induced diabetes and two suramin treatments (10 mg/kg, ip). Vascular endothelial growth factor A expression was evaluated by western blot of glomeruli and immunofluorescence of the renal cortex. RT-PCR for receptors Vegfr1 mRNA and Vegfr2 mRNA quantitation was performed. The soluble adhesive molecules (sICAM-1, sVCAM-1) in blood were measured by ELISA and the vasoreactivity of interlobar arteries to acetylcholine was evaluated using wire myography. Suramin administration reduced the expression and intraglomerular localisation of VEGF-A. Increased VEGFR-2 expression in diabetes was reduced by suramin to non-diabetic levels. Diabetes reduced the sVCAM-1 concentrations. Suramin in diabetes restored acetylcholine relaxation properties to non-diabetic levels. In conclusion, suramin affects the renal VEGF-A/VEGF receptors axis and has a beneficial impact on endothelium-dependent relaxation of renal arteries. Thus, suramin may be used as a pharmacological agent to investigate the potential role of VEGF-A in the pathogenesis of renal vascular complications in short-term diabetes.