Merck
  • Microwave assisted synthesis of some hybrid molecules derived from norfloxacin and investigation of their biological activities.
Microwave assisted synthesis of some hybrid molecules derived from norfloxacin and investigation of their biological activities.
European journal of medicinal chemistry (2013-07-23)
Meltem Yolal Mentese, Hacer Bayrak, Yıldız Uygun, Arif Mermer, Serdar Ulker, Sengul Alpay Karaoglu, Neslihan Demirbas
ABSTRACT
Norfloxacin was converted to 7-(4-amino-2-fluorophenyl)piperazin derivative (2) via the formation of nitro compound. The synthesis of the norfloxacin derivatives containing 1,3-thiazole or 1,3-thiazolidin moiety was performed from the reaction of 4-chlorophenacylbromide or ethyl bromoacetate with compounds 4-7 obtained starting from 2. 3-Fluoro-4-[4-(2-methoxyphenyl)piperazin-1-yl]aniline (14), 5-{[4-(2-methoxyphenyl)piperazin-1-yl]methyl}-4-phenyl-4H-1,2,4-triazole-3-thiol (18) and {[4-(2-methoxy phenyl)piperazin-1-yl]methyl}-1,3,4-oxadiazol-2-thiol (19) were obtained starting from 1-(2-methoxyphenyl)piperazine by several steps. The treatment of hydrazide (16) with several aldehydes afforded N'-[(2-hydroxyphenyl)methylen]- (20), N'-[(3-hydroxy-4-methoxy phenyl)methylen]- (21) or N'-[1H-indol-3-ylmethylene]-2-[4-(2-methoxyphenyl)piperazin-1-yl]acetohydrazide (22). Then, compounds 14, 18, 19 and 22 were condensed with 7-[4-(chloroacetyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid (3) that was obtained from norfloxacine. All newly synthesized compounds were screened for their antimicrobial activities and some of them exhibited excellent activity. Moreover, one compound was found to have antiurease activity.
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, ~1 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type IX, powder, 50,000-100,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean)
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, white, ~8 U/mg