Merck
  • Reversibility of structural changes in the resistance vessels in hypertension: a review of studies with pindolol.
Reversibility of structural changes in the resistance vessels in hypertension: a review of studies with pindolol.
British journal of clinical pharmacology (1987-01-01)
H Duner
ABSTRACT
1. In recent years evidence has accumulated which indicates that although various beta-adrenoceptor antagonists are equally effective in lowering blood pressure in patients with essential hypertension the mechanisms of action of the different drugs are heterogeneous. 2. Pindolol, a beta-adrenoceptor antagonist with pronounced intrinsic sympathomimetic activity (ISA) appears, to some extent, to act via peripheral vascular mechanisms. 3. Following prolonged treatment with pindolol in essential hypertension peripheral vascular resistance at maximal vasodilatation has been found to be decreased compared with the pretreatment values suggesting that the structural vascular changes characteristic of established hypertension may be reversible.
Product Description
Pindolol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder