Skip to Content
Merck
pH standard solution pH 10.01 (25 °C)

pH standard solution

pH standard solution pH 10.01 (25 °C)

Products

Product Category
Brand
pH Value
Agency
Format
Quality Segment
Application
Reference Material Type