Merck
  • Simple chiral diene ligands provide high enantioselectivities in transition-metal-catalyzed conjugate addition reactions.
Simple chiral diene ligands provide high enantioselectivities in transition-metal-catalyzed conjugate addition reactions.
Organic letters (2008-09-06)
Kazuhiro Okamoto, Tamio Hayashi, Viresh H Rawal
ABSTRACT
Chiral dienes possessing the bicyclo[2.2.2]octadiene framework were prepared readily through the [4 + 2] cycloaddition of ( R)-alpha-phellandrene with methyl propiolate as the key step. Diene 9, substituted with a tertiary alcohol on one of the two double bonds, is prepared in just one step from the cycloadduct and is highly effective as a chiral ligand for rhodium-catalyzed asymmetric conjugate addition reactions, giving the corresponding addition products with higher enantioselectivity than other chiral dienes.
Sigma-Aldrich
(R)-(−)-α-Phellandrene, ≥95.0% (sum of enantiomers, GC)