Merck
  • Identification and characterization of a hyperthermophilic GH9 cellulase from the Arctic Mid-Ocean Ridge vent field.
Identification and characterization of a hyperthermophilic GH9 cellulase from the Arctic Mid-Ocean Ridge vent field.
PloS one (2019-09-07)
Anton A Stepnov, Lasse Fredriksen, Ida H Steen, Runar Stokke, Vincent G H Eijsink
ABSTRACT
A novel GH9 cellulase (AMOR_GH9A) was discovered by sequence-based mining of a unique metagenomic dataset collected at the Jan Mayen hydrothermal vent field. AMOR_GH9A comprises a signal peptide, a catalytic domain and a CBM3 cellulose-binding module. AMOR_GH9A is an exceptionally stable enzyme with a temperature optimum around 100°C and an apparent melting temperature of 105°C. The novel cellulase retains 64% of its activity after 4 hours of incubation at 95°C. The closest characterized homolog of AMOR_GH9A is TfCel9A, a processive endocellulase from the model thermophilic bacterium Thermobifida fusca (64.2% sequence identity). Direct comparison of AMOR_GH9A and TfCel9A revealed that AMOR_GH9A possesses higher activity on soluble and amorphous substrates (phosphoric acid swollen cellulose, konjac glucomannan) and has an ability to hydrolyse xylan that is lacking in TfCel9A.
Sigma-Aldrich
2,4-Dihydroxybenzophenone, 99%