Merck
  • The effect of ractopamine hydrochloride on the carcass cutting yields of finishing swine.
The effect of ractopamine hydrochloride on the carcass cutting yields of finishing swine.
Journal of animal science (1991-08-01)
C R Stites, F K McKeith, S D Singh, P J Bechtel, D H Mowrey, D J Jones
ABSTRACT
This study was designed to evaluate growth performance, carcass cutting yield, and processing characteristics of boneless hams and bellies from finishing pigs fed diets containing 0, 5, 10, or 20 ppm of the phenethanolamine ractopamine hydrochloride (RAC). Sixty pigs were blocked by starting weight and randomly assigned to pens (four pigs/pen) within each of three blocks. Treatments were then randomly assigned to the pens to total six pens of the 0-ppm level and three pens each at the 5-, 10-, and 20-ppm RAC levels. Weight gain and feed consumption were monitored and animals were slaughtered by weight block after approximately 48 d on trial. Slaughter weight, ADG, and feed/gain were improved (P less than .05) for RAC treatments. Dressing percentage was higher and increased linearly (P less than .05) for RAC treatments. Carcass weight, length, leaf fat weight, backfat thickness, loin eye area, and color, marbling, and firmness of the longissimus were evaluated. The RAC-treated carcasses were heavier (P less than .05) and loin eye area increased linearly (P less than .05). One side of each carcass was fabricated using National Association of Meat Purveyors specifications. Trimmed hams and loins from the RAC treatments were heavier (P less than .05) than those from control animals. No differences (P greater than .05) in carcass cutting yield (percentage of trimmed primal cuts) were observed between treatments. However, trimmed hams and loins from the 20-ppm RAC treatment represented a greater (P less than .05) percentage of carcass weight than did those from control animals. Ractopamine did not affect raw belly or bacon characteristics (P less than .05).(ABSTRACT TRUNCATED AT 250 WORDS)
Product Description
Supelco
Ractopamine hydrochloride, VETRANAL®, analytical standard