Merck
  • Noninvasive monitoring of HPMA copolymer-RGDfK conjugates by magnetic resonance imaging.
Noninvasive monitoring of HPMA copolymer-RGDfK conjugates by magnetic resonance imaging.
Pharmaceutical research (2009-01-23)
Bahar Zarabi, Mark P Borgman, Jiachen Zhuo, Rao Gullapalli, Hamidreza Ghandehari
ABSTRACT
To evaluate the tumor targeting potential of N-(2-hydroxypropyl)methacrylamide (HPMA) copolymer-gadolinium(Gd)-RGDfK conjugates by magnetic resonance (MR) T1-mapping. HPMA copolymers with and without RGDfK were synthesized to incorporate side chains for Gd chelation. The conjugates were characterized by their side-chain contents and r(1) relaxivity. In vitro integrin-binding affinities of polymeric conjugates were assessed via competitive cell binding assays on HUVEC endothelial cells and MDA-MB-231 breast cancer cells. In vivo MR imaging was performed on MDA-MB-231 tumor-bearing SCID mice at different time points using non-targetable and targetable polymers. The specificity of alphavbeta3 targeting was assessed by using non-paramagnetic targetable polymer to block alphavbeta3 integrins followed by injection of paramagnetic targetable polymers after 2 h. The polymer conjugates showed relaxivities higher than Gd-DOTA. Endothelial cell binding studies showed that IC(50) values for the copolymer with RGDfK binding to alphavbeta3 integrin-positive HUVEC and MDA-MB-231 cells were similar to that of free peptide. Significantly lower T1 values were observed at the tumor site after 2 h using targetable conjugate (p < 0.012). In vivo blocking study showed significantly higher T1 values (p < 0.045) compared to targetable conjugate. These results demonstrate the potential of this conjugate as an effective targetable MR contrast agent for tumor imaging and therapy monitoring.
Sigma-Aldrich
Hexamethylphosphoramide, purum, ≥98.0% (GC)
Sigma-Aldrich
Hexamethylphosphoramide, absolute, over molecular sieve (H2O ≤0.03%), ≥98.0% (GC)
Sigma-Aldrich
Hexamethylphosphoramide, 99%