Merck
Murine macrophages response to iron.
Journal of proteomics (2012-07-28)
Rita Polati, Annalisa Castagna, Alessandra Maria Bossi, Tiziana Alberio, Ivana De Domenico, Jerry Kaplan, Anna Maria Timperio, Lello Zolla, Federica Gevi, Angelo D'Alessandro, Ryan Brunch, Oliviero Olivieri, Domenico Girelli
ABSTRACT
Macrophages play a critical role at the crossroad between iron metabolism and immunity, being able to store and recycle iron derived from the phagocytosis of senescent erythrocytes. The way by which macrophages manage non-heme iron at physiological concentration is still not fully understood. We investigated protein changes in mouse bone marrow macrophages incubated with ferric ammonium citrate (FAC 10 μM iron). Differentially expressed spots were identified by nano RP-HPLC-ESI-MS/MS. Transcriptomic, metabolomics and western immunoblotting analyses complemented the proteomic approach. Pattern analysis was also used for identifying networks of proteins involved in iron homeostasis. FAC treatment resulted in higher abundance of several proteins including ferritins, cytoskeleton related proteins, glyceraldehyde-3-phosphate dehydrogenase (GAPDH) at the membrane level, vimentin, arginase, galectin-3 and macrophage migration inhibitory factor (MIF). Interestingly, GAPDH has been recently proposed to act as an alternative transferrin receptor for iron acquisition through internalization of the GAPDH-transferrin complex into the early endosomes. FAC treatment also induced the up-regulation of oxidative stress-related proteins (PRDX), which was further confirmed at the metabolic level (increase in GSSG, 8-isoprostane and pentose phosphate pathway intermediates) through mass spectrometry-based targeted metabolomics approaches. This study represents an example of the potential usefulness of "integarated omics" in the field of iron biology, especially for the elucidation of the molecular mechanisms controlling iron homeostasis in normal and disease conditions. This article is part of a Special Issue entitled: Integrated omics.
Sigma-Aldrich
Ammonium iron(III) citrate, CP, green
Sigma-Aldrich
Ammonium iron(III) citrate, reagent grade, powder
Millipore
Fraser Supplement, suitable for microbiology