Merck
  • Hydrochlorothiazide versus chlorthalidone: evidence supporting their interchangeability.
Hydrochlorothiazide versus chlorthalidone: evidence supporting their interchangeability.
Hypertension (Dallas, Tex. : 1979) (2003-11-26)
Barry L Carter, Michael E Ernst, Jerome D Cohen
ABSTRACT
Thiazide diuretics are one of the preferred pharmacologic treatments for hypertension. Hydrochlorothiazide and chlorthalidone have been the 2 most commonly used diuretics in major clinical trials. Treatment guidelines and compendia often consider these 2 drugs interchangeable agents within the class of thiazide or thiazide-like diuretics. Many sources list them as equipotent. Despite these beliefs, there is some suggestion that cardiovascular outcomes are not necessarily the same with these 2 drugs. We conducted a literature search from 1960 to 2003 to identify studies that evaluated the pharmacokinetic and blood pressure-lowering effects of these 2 agents. There are significant pharmacokinetic and pharmacodynamic differences between these diuretics. Chlorthalidone is approximately 1.5 to 2.0 times as potent as hydrochlorothiazide, and the former has a much longer duration of action. Whether these pharmacokinetic and pharmacodynamic features cause differences in outcomes is not known.
Sigma-Aldrich
Chlorthalidone, ≥98% (HPLC)
USP
Chlorthalidone, United States Pharmacopeia (USP) Reference Standard
Chlorthalidone, European Pharmacopoeia (EP) Reference Standard