Merck
  • Oxcarbazepine versus carbamazepine monotherapy for partial onset seizures.
Oxcarbazepine versus carbamazepine monotherapy for partial onset seizures.
The Cochrane database of systematic reviews (2009-10-13)
Marcus W Koch, Susanne Kl Polman
ABSTRACT
Partial onset seizures are often treated with the standard antiepileptic drug carbamazepine. Oxcarbazepine is a newer antiepileptic drug related to carbamazepine that is claimed to be better tolerated. To compare efficacy and tolerability of carbamazepine and oxcarbazepine monotherapy for partial onset seizures. We searched the Cochrane Epilepsy Group Specialised Register (4 August 2009), the Cochrane Central Register of Controlled Trials (CENTRAL, The Cochrane Library issue 3, 2009), MEDLINE (January 1966 to May 2008), reference lists of relevant articles and conference proceedings. We also contacted manufacturers and researchers in the field for published or unpublished data. Blinded and unblinded randomised controlled trials of carbamazepine versus oxcarbazepine monotherapy for partial onset seizures. Both authors independently assessed trial quality, according to the guidelines in the Cochrane Reviewer's Handbook, and extracted information about study population, type of intervention, outcome measures and study design. All analyses in this review are by intention-to-treat. We tested for statistical heterogeneity among the identified studies using the chi-squared test. Three trials (723 participants) were included. Only one trial used adequate outcome measures of efficacy; therefore, the results pertaining to efficacy are based on a single trial, whereas the results pertaining to adverse events are based on all three included trials. There was no overall difference in time to treatment withdrawal between the two drugs (hazard ratio (HR) of oxcarbazepine (OXC) versus carbamazepine (CBZ): 1.04, 95% confidence interval (CI) 0.78 to 1.39). Further analyses showed no significant difference in treatment withdrawal for unacceptable side effects (HR of OXC versus CBZ: 0.85, 95% CI 0.59 to 1.24) and in treatment withdrawal for inadequate seizure control (HR of OXC versus CBZ: 1.33, 95% CI 0.82 to 2.15). Oxcarbazepine and carbamazepine appeared to be similarly effective and well tolerated although the confidence intervals around estimates were wide and do not rule out the possibility of important differences existing. Significantly fewer patients on carbamazepine treatment developed nausea or vomiting, or both (odds ratio of OXC versus CBZ: 3.15, 95% CI 1.39 to 7.14). Oxcarbazepine and carbamazepine appear to be similarly effective and well tolerated. However, the possibility of important differences existing between these drugs cannot be ruled out.
Product Description
Supelco
Oxcarbazepine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Carbamazepine, analytical standard
Oxcarbazepine, European Pharmacopoeia (EP) Reference Standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
USP
Oxcarbazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Oxcarbazepine, ≥98% (HPLC), solid
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®