Merck
  • Intracellular ATP-binding cassette transporter A3 is expressed in lung cancer cells and modulates susceptibility to cisplatin and paclitaxel.
Intracellular ATP-binding cassette transporter A3 is expressed in lung cancer cells and modulates susceptibility to cisplatin and paclitaxel.
Oncology (2013-05-22)
Tobias R Overbeck, Timo Hupfeld, Doris Krause, Regina Waldmann-Beushausen, Bjoern Chapuy, Bjoern Güldenzoph, Thiha Aung, Nobuya Inagaki, Friedrich A Schöndube, Bernhard C Danner, Lorenz Truemper, Gerald G Wulf
ABSTRACT
Patients with advanced-stage bronchial cancer benefit from systemic cytostatic therapy, in particular from regimens integrating cisplatin and taxanes. However, eventual disease progression leads to a fatal outcome in most cases, originating from tumor cells resisting chemotherapy. We here show that the intracellular ATP-binding cassette transporter A3 (ABCA3), previously recognized as critical for the secretion of surfactant components from type 2 pneumocytes, is expressed in non-small-cell lung cancer (NSCLC) cells. With some heterogeneity in a given specimen, expression levels detected immunohistochemically in primary cancer tissue were highest in adenocarcinomas and lowest in small cell lung cancers. Genetic silencing of ABCA3 in the NSCLC cell line models A549, NCI-H1650 and NCI-H1975 significantly increased tumor cell susceptibility to the cytostatic effects of both cisplatin (in all cell lines) and paclitaxel (in two of three cell lines). Taken together, ABCA3 emerges as a modulator of NSCLC cell susceptibility to cytostatic therapy.
Sigma-Aldrich
Anti-ABCA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.