Merck
  • Maternal perchlorate levels in women with borderline thyroid function during pregnancy and the cognitive development of their offspring: data from the Controlled Antenatal Thyroid Study.
Maternal perchlorate levels in women with borderline thyroid function during pregnancy and the cognitive development of their offspring: data from the Controlled Antenatal Thyroid Study.
The Journal of clinical endocrinology and metabolism (2014-07-25)
Peter N Taylor, Onyebuchi E Okosieme, Rhian Murphy, Charlotte Hales, Elisabetta Chiusano, Aldo Maina, Mohamed Joomun, Jonathan P Bestwick, Peter Smyth, Ruth Paradice, Sue Channon, Lewis E Braverman, Colin M Dayan, John H Lazarus, Elizabeth N Pearce
ABSTRACT
Thyroid dysfunction is associated with impaired cognitive development. Perchlorate decreases thyroidal iodine uptake, potentially reducing thyroid hormone production. It is unclear whether perchlorate exposure in early life affects neurodevelopment. Historical cohort analysis. From 2002 to 2006, 21,846 women at gestational age <16 weeks recruited from antenatal clinics in Cardiff, UK and Turin, Italy were enrolled in the Controlled Antenatal Thyroid Screening Study (CATS). We undertook a retrospective analysis of 487 mother-child pairs in mothers who were hypothyroid/hypothyroxinemic during pregnancy and analyzed whether first trimester maternal perchlorate levels in the highest 10% of the study population were associated with increased odds of offspring IQ being in the lowest 10% at 3 years of age. Maternal urinary perchlorate, offspring IQ. Urine perchlorate was detectable in all women (median 2.58 μg/L); iodine levels were low (median 72 μg/L). Maternal perchlorate levels in the highest 10% of the population increased the odds of offspring IQ being in the lowest 10% OR = 3.14 (95% CI 1.38, 7.13) P = .006 with a greater negative impact observed on verbal OR = 3.14 (95% CI 1.42, 6.90) P = .005 than performance IQ. Maternal levothyroxine therapy did not reduce the negative impact of perchlorate on offspring IQ. This is the first study using individual-level patient data to study maternal perchlorate exposure and offspring neurodevelopment and suggests that high-end maternal perchlorate levels in hypothyroid/hypothyroxinemic pregnant women have an adverse effect on offspring cognitive development, not affected by maternal levothyroxine therapy. These results require replication in additional studies, including in the euthyroid population.
Sigma-Aldrich
Iodine, SAJ first grade, ≥99.0%
Sigma-Aldrich
Iodine, JIS special grade, ≥99.8%
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)