Skip to Content
Merck
All Photos(1)

Key Documents

E0637

Sigma-Aldrich

β-Endorphin bovine, camel, ovine

≥97% (HPLC)

Synonym(s):

β-Lipotropin 61-91

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C155H250N42O44S
CAS Number:
Molecular Weight:
3437.96
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H]2CCCN2C(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](CCSC)NC(=O)[C@H](Cc3ccccc3)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc4ccc(O)cc4)[C@@H](C)O)[C@@H](C)O)C(C)C)[C@@H](C)O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc5cnc[nH]5)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C155H250N42O44S/c1-17-82(9)123(150(235)182-99(44-29-34-61-160)134(219)186-109(70-116(164)206)139(224)171-84(11)128(213)183-108(69-92-72-166-78-170-92)144(229)178-96(41-26-31-58-157)132(217)175-95(40-25-30-57-156)131(216)169-75-120(210)173-103(155(240)241)51-54-115(163)205)193-151(236)124(83(10)18-2)192-129(214)85(12)172-140(225)110(71-117(165)207)185-133(218)97(42-27-32-59-158)177-143(228)107(68-90-38-23-20-24-39-90)184-141(226)104(64-79(3)4)188-152(237)126(87(14)201)195-149(234)122(81(7)8)191-145(230)105(65-80(5)6)187-148(233)113-45-35-62-197(113)154(239)127(88(15)202)196-137(222)100(50-53-114(162)204)179-146(231)111(76-198)189-135(220)98(43-28-33-60-159)176-136(221)101(52-55-121(211)212)180-147(232)112(77-199)190-153(238)125(86(13)200)194-138(223)102(56-63-242-16)181-142(227)106(67-89-36-21-19-22-37-89)174-119(209)74-167-118(208)73-168-130(215)94(161)66-91-46-48-93(203)49-47-91/h19-24,36-39,46-49,72,78-88,94-113,122-127,198-203H,17-18,25-35,40-45,50-71,73-77,156-161H2,1-16H3,(H2,162,204)(H2,163,205)(H2,164,206)(H2,165,207)(H,166,170)(H,167,208)(H,168,215)(H,169,216)(H,171,224)(H,172,225)(H,173,210)(H,174,209)(H,175,217)(H,176,221)(H,177,228)(H,178,229)(H,179,231)(H,180,232)(H,181,227)(H,182,235)(H,183,213)(H,184,226)(H,185,218)(H,186,219)(H,187,233)(H,188,237)(H,189,220)(H,190,238)(H,191,230)(H,192,214)(H,193,236)(H,194,223)(H,195,234)(H,196,222)(H,211,212)(H,240,241)/t82-,83-,84-,85-,86+,87+,88+,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,122-,123-,124-,125-,126-,127-/m0/s1

InChI key

HQALISCIMNBRAE-XJTHNYJFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-His-Lys-Lys-Gly-Gln

Biochem/physiol Actions

Potent endogenous opioid protein that is derived from propiomelanocortin, a protein found in the brain, anterior pituitary, skin, immune system, and other peripheral sites. It is released in response to painful stimuli and has potent antinociceptive activity that is mediated through its action on μ receptors in brain and by μ and κ receptors in the spinal cord.
Potent endogenous opioid protein; released in response to painful stimuli and has potent antinociceptive activity.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service