Skip to Content
Merck
All Photos(1)

Key Documents

F5022

Sigma-Aldrich

Fibronectin-like Protein Polymer genetically engineered

lyophilized powder, autoclaved, BioReagent, suitable for cell culture

Synonym(s):

ProNectin® F

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

sterility

autoclaved

product line

BioReagent

form

lyophilized powder

mol wt

75 kDa by gene sequence (differences are due to the unusual amino acid composition of the protein)
~110 kDa by SDS-PAGE

packaging

pkg of 1 mg
pkg of 5 mg

technique(s)

cell culture | mammalian: suitable

solubility

4.5  M LiClO4: 1 mg/mL (stable for 12 mo. at RT.)

shipped in

dry ice

storage temp.

room temp

Application

Recommended as a cell culture substratum at 2-10 μg/cm2.

Packaging

Supplied with diluent in separate vial.

Other Notes

Positively-charged, protein polymer which incorporates multiple copies of the RGD cell attachment epitope from human fibronectin between repeated structural peptide units.

Reconstitution

Dilutions can be made to a working concentration with balanced salt solution; stable up to 2 days at RT. Coated labware is stable at least 4 mos at RT.

Legal Information

ProNectin is a registered trademark of Sanyo Chemical Industries, Ltd

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Receptor-specific serum-free cell attachment using a highly stable engineered protein polymer.
Esty, A.
American Biotechnology Laboratory, 44-44 (1991)
Polly A Machin et al.
Frontiers in immunology, 14, 1180886-1180886 (2023-06-29)
Rac-GTPases and their Rac-GEF activators play important roles in neutrophil-mediated host defence. These proteins control the adhesion molecules and cytoskeletal dynamics required for neutrophil recruitment to inflamed and infected organs, and the neutrophil effector responses that kill pathogens. Here, we
Zoltán Jakus et al.
The Journal of experimental medicine, 206(3), 577-593 (2009-03-11)
beta(2) integrins and Fcgamma receptors are critically involved in neutrophil activation at the site of inflammation. Both receptor types trigger a receptor-proximal tyrosine phosphorylation cascade through Src family kinases and Syk, but further downstream signaling events are poorly understood. We
Lamyaa Shaban et al.
PLoS pathogens, 16(5), e1008576-e1008576 (2020-05-12)
Yersinia suppress neutrophil responses by using a type 3 secretion system (T3SS) to inject 6-7 Yersinia effector proteins (Yops) effectors into their cytoplasm. YopH is a tyrosine phosphatase that causes dephosphorylation of the adaptor protein SKAP2, among other targets in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service