
3-Amino-2,4,6-triiodobenzoic acid

Linear Formula:
H2NC6HI3CO2H
CAS Number:
Molecular Weight:
514.83
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing