Skip to Content
Merck
Benzo[e]pyrene BCR®, certified reference material

Benzo[e]pyrene

Benzo[e]pyrene BCR®, certified reference material
Synonym(s):
1,2-Benzpyrene, 4,5-Benzpyrene
Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing