Skip to Content
Merck
Ethylparaben certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Ethylparaben

Ethylparaben certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Ethyl 4-hydroxybenzoate, Ethylparaben
Linear Formula:
HOC6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique